Geldanamycin ≥98%

Supplier: Ambeed
Warning

A164935-1MG A164935-5MG A164935-100MG A164935-10MG A164935-250MG
77667-308EA 15 USD
77667-308 77667-310 77879-105 77879-106 77879-107
Geldanamycin ≥98%
Geldanamycin

Caution: for Research Use Only

Formula: C₂₉H₄₀N₂O₉
MDL Number: MFCD00274570
CAS Number: 30562-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR