Posaconazole 98%

Supplier: Ambeed
Warning

A133379-10G A133379-10MG A133379-1G A133379-50MG A133379-5G A133379-25G A133379-100G A133379-100MG A133379-250MG A133379-5MG
76719-022EA 219 USD
76719-022 76719-024 76719-026 76719-028 76719-030 77592-534 77599-910 77599-912 77599-914 77666-914
Posaconazole 98%
Posaconazole

Caution: for Research Use Only

Formula: C₃₇H₄₂F₂N₈O₄
MDL Number: MFCD00941162
CAS Number: 171228-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR