Titanium Forceps with Tying Platform, Straight
Supplier: WORLD PRECISION INSTRUMENTS LLC
WP500097
77666-290EA 220 USD
77666-290
Titanium Forceps with Tying Platform, Straight
Forceps
Forceps with a tying platform are designed to grasp, and manipulate tissues or objects during surgical procedures. They flat tying platform is near the tips where suture can be tied. This gives you a stable area for suturing or ligating.
  • 100% non-corrosive (highly suitable for seawater procedures)
  • 100% non-magnetic (MRI compatible)
  • 40% lighter than stainless steel (reduces hand fatigue)
  • Forceps' tips are coated with tungsten carbide for increased gripping power
  • Anodized, non-glare blue finish
The slender handles are textured for a firm grip, and the hole patterns in the handle makes the instrument incredibly lightweight.