Student Micro Surgery Scissors
Supplier: WORLD PRECISION INSTRUMENTS LLC
WP28105S
77665-670EA 84 USD
77665-670
Student Micro Surgery Scissors
Scissors
Our economy line of spring scissors are perfect for students who are doing small animal surgeries or dissection in a research laboratory. Student spring scissors are manufactured from hardened surgical grade stainless steel. They are perfect for classroom use or as disposable instruments.
  • 150 mm long
  • 4 mm cuting edge