One Channel Free Radical Analyzer, World Precision Instruments
Supplier: WORLD PRECISION INSTRUMENTS LLC
TBR1025
77665-210EA 5600 USD
77665-210
One Channel Free Radical Analyzer, World Precision Instruments
Analyzers Elemental Analysers
Fast, reliable, real-time detection - measure redox-reactive species. Real-time detection and measurement of a variety of redox-reactive species is fast and easy using the electrochemical (amperometric) detection principle employed in the TBR4100.
  • Real-time detection using electrochemical microsensors
  • Integrated system includes one temperature sensor, your choice of one additional sensor and a start-up kit
  • Current measurement range from 300 fA to 10 µA (four ranges) permits wide dynamic range for detection
  • Wide bandwidth allows recording of fast events
  • Measure carbon monoxide from 10 nM to 10 µM
  • Measure nitric oxide from <0.3 nM to 100 µM
  • Measure hydrogen peroxide <10 nM to 100 mM
  • Measure hydrogen sulfide
  • Measure glucose
  • Measure oxygen from 0.1 to 100%
  • Isolated architecture allows Lab-Trax interface to simultaneously measure free radical and independent analog data (for example, ECG, BP, etc.) on any channel
  • Single channel free radical detection
This optically isolated four-channel free radical analyzer has ultra low noise and independently operated channels. Applications: Free radical detection (NO, H₂O₂, H₂S, CO, O₂ and glucose)
Benefits like measure up to four different species and temperature in the same preparation or simultaneous measurement in four different preparations. Lab-Trax data acquisition system is flexible.