Omega-Tip-Z™ Electrode Impedance Meter
Supplier: WORLD PRECISION INSTRUMENTS LLC
SYS-OMEGAZ
77665-192EA 3662.63 USD
77665-192
Omega-Tip-Z™ Electrode Impedance Meter
Electromagnetic Field Meters
Omega-Tip-Z™ was created especially for measuring impedance in etched tung­sten, platinum-iridium* and steel microelectrodes, as well as electrolyte-filled micropipettes. The meter's AC impedance-measuring circuit is unaffected by electrode offset or tip junction potentials.
  • Battery operated
  • Portable
  • Measures mV and MΩ impedance of metal or glass microelectrodes
  • Low noise probe included
  • Determine resistance of glass capillary electrode during micropipette beveling process.
  • Measure impedance of metal microelectrodes
The gold-plated miniature probe lets you conveniently monitor micro-electrode impedance in electrolytes, and an electrode tip cleaning feature lets you remove buildup quickly. Omega-Tip-Z can also measure DC electrode tip potentials up to 2000 millivolts. The instrument operates for hundreds of hours without battery failure.
Applications: Impedance measurements provide valuable information about the electrode-electrolyte interface, including the resistance and capacitance of the system.