Rodent Brain Matrices, Acrylic
Supplier: WORLD PRECISION INSTRUMENTS LLC
RBMA-200C RBMA-200S RBMA-300C RBMA-300S RBMA-600C RBMA-600S
77665-076EA 197 USD
77665-076 77665-078 77665-080 77665-082 77665-084 77665-086
Rodent Brain Matrices, Acrylic
Specimens Preserved Specimens Preserved Animal Materials
Using a WPI brain matrix, you can get thin and uniform brain slices, which is crucial for histological analysis, immunohistochemistry, molecular biology, and more. This consistency is essential for accurate comparisons between different brains and experimental conditions. These WPI rodent brain matrices are made of acrylic, a durable material. They have precision-cut channels or slots that hold the brain in place during slicing. Choose your brain matrix based on your species (mouse or rat) and the size of the animal.
  • Acrylic brain matrices for mouse and rat
  • Sagittal or coronal slices
  • Repeatable sections, 1 mm spacing
  • Can be chilled and sterilized
  • Olfactory/spinal/notch cut into every matrix
The WPI brain matrices are designed for sagittal or coronal slices. These brain matrices give you consistent slices that are 1 mm thick, and the olfactory spinal notch is cut into every brain matrix. Get standardized and precise brain sectioning. Use WPI brain matrices to ensure the reproducibility and accuracy of your experiments for a better understanding of the structure and function of the rodent brain.