Platinum/Iridium Wires, Teflon Coated
Supplier: WORLD PRECISION INSTRUMENTS LLC
PTT0110 PTT0203 PTT0502
77665-038EA 358 USD
77665-038 77665-040 77665-042
Platinum/Iridium Wires, Teflon Coated
Wire
Platinum/iridium wire Teflon-coated is an alloy of 90% platinum and 10% iridium, giving excellent tensile strength and corrosion resistance.
  • Available in 36, 44 and 50 AWG
  • Available in precut lengths
The Teflon coating is 4 μm (150 micro-in.) thick. The Teflon coating is designed to reduce surface friction, only. It is not insulation.