SMARTouch™ Microinjection Syringe Pump Controller
Supplier: WORLD PRECISION INSTRUMENTS LLC
MICRO2T
77664-816EA 3524.61 USD
77664-816
SMARTouch™ Microinjection Syringe Pump Controller
Controllers Utility Controllers
The touch-screen display on WPI's MICRO2T SMARTouch™ controller offers an intuitive, user-friendly interface. Our MICRO2T allows for simultaneous control of the UMP3 syringe pump.
  • Splash-proof touch screen
  • Built-in memory configured for saving settings between use
  • A dual mode motor drive
The MICRO2T SMARTouch™ controller for the UMP3 features patent pending technology which includes:
Total system calibration - Calibrate the syringe and the controller together as a system. This feature eliminates variability of the syringe and delivers (or withdraws) the configured volume.
Smart smoothness - The controller can be set to automatically adjust microstepping according to the injection rate to deliver (or withdraw) smoothly.
User defined travel limits - Set the limits for your NanoSyringe on your MICRO2T. This prevents your UMP3 from exceeding its drive range.
The MICRO2T SMARTouch™ controller has a graphical indication of the rate and volume remaining in your NanoSyringe. It offers end-stop detection that is dependent on the syringe volume. You can control two pumps independently the MICRO2T's dual capability. The controller also has automatic pump detection and a Pause/Resume feature for incremental dosing during infusion, or sample collection upon withdraw. The MICRO2T SMARTouch™ controller is fully compatible with all previous models of the UMP3.