Microelectrode Holders, MEH3SW
Supplier: WORLD PRECISION INSTRUMENTS LLC
MEH3SW10 MEH3SW12 MEH3SW15 MEH3SW20
77664-728EA 82 USD
77664-728 77664-730 77664-732 77664-734
Microelectrode Holders, MEH3SW
Electrode Holders
Microelectrode holder-half-cells couple fluid-filled glass micropipettes to high input impedance amplifiers. All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters.
  • Male connector
  • Wire half-cell
  • No pressure port
  • Screw cap
A Ag/AgCl pellet (or a silver wire) molded into the holder body provides stable potential. Electrical connection is made via male 2 mm pins or female 2 mm sockets. The pipette may be mounted axially or at right angles to the holder. Pipettes are held with screw-caps or rubberMore Product Information