Manual Micromanipulator, M3301
Supplier: WORLD PRECISION INSTRUMENTS LLC
M3301L M3301-M3-L M3301R
77664-578EA 1700 USD
77664-578 77664-580 77664-584
Manual Micromanipulator, M3301
Micromanipulators
The M3301 is a popular Manual Micromanipulator, because it is accurate, lightweight and well-engineered. This solid manipulator outsells all others worldwide for high precision experiments where magnification is in the range of up to 250×.
  • Lightweight (550 g)
  • Sure, repeatable movement without drift
  • Choice of optional M-3 tilting base
  • Thumbscrew included for metric anti-vibration platforms with M6 holes
  • 5464 Base Weight and 15873 Angled Electrode Holder sold separately
  • Comes installed with a 12 mm clamp for the M10 magnetic base
It has a slim, space-saving design. Units can stand tightly grouped, since all control knobs project to the rear. Because the control knobs are clustered within an 8 cm area in a single vertical plane, resolution is quick. The hand works blindly while the eye monitors the microscopic image. Vernier scales allow readings to 0.1 mm. X-axis fine control allows readings to 10 μm.
The rack-and-pinion drive, V-shaped guideways and cross roller bearings ensure smooth movement that is sure and repeatable, without drift, side play, backlash or sticking. Contact parts are milled of hardened steel for high performance and long life.