Light Source Assay, FO-6000
Supplier: WORLD PRECISION INSTRUMENTS LLC
FO-6000
77664-340EA 4157.58 USD
77664-340
Light Source Assay, FO-6000
Lights Spectrum Lamps
The FO-6000 is a continuous fiber optic light source featuring an extended visible part of the light source (380 to 1700 nm). It has a SMA fiber optic connector. Both, shutter and lamp can be controlled via a switch or external TTL triggering. This light source offers a wide assortment of applications. A special feature of the FO-6000 is its color balancing optics, which shifts the usable range of the light source from traditional 420 nm down to 380 nm wavelength. Due to its thermally controlled optical bench, it is particularly suitable for low noise and low drift applications.
  • Visible light source and high light output
  • 380 to 1700 nm
  • Low drift <0.5 mAU/h
  • SMA fiber connection
  • Electrical shutter control (switch and TTL)
  • Compact - lightweight and ow power consumption
  • External triggering
  • 15 to 20 minutes warming up time
  • Negligible heat generation
Benefits:
Bulb with 10000 h lifetime
Fits all of WPI's fiber optic probes and flow cells
Design matched to LWCC applications
Temperature controlled optical bench
Applications:
Low noise VIS absorbance measurements
Trace analysis of nutrients in seawater and freshwater
CDOM analysis in seawater
Analytical chemistry, environmental science and life science