Electrode Voltage
Supplier: WORLD PRECISION INSTRUMENTS LLC
EKV
77664-244EA 115 USD
77664-244
Electrode Voltage
Electrodes Other Electrodes
Current (red) and voltage (black) electrodes are threaded and screw securely into the end of each Luer tipped cartridge. The Luer tip then plugs securely into the Luer openings of the chamber. The cable from each electrode terminates with a 2 mm pin which may be plugged into most voltage/current clamps.
  • Ussing chamber current electrode
  • Works with most voltage/current clamps
The miniature electrode gel cartridge is a small plastic tube with a male Luer tip identical to those at the tip of hypodermic syringes. The tube may be filled with different gel materials. Agar is commonly used, but other gel materials may also be satisfactory.