Electrode Kit USC
Supplier: WORLD PRECISION INSTRUMENTS LLC
EK1
77664-238EA 499 USD
77664-238
Electrode Kit USC
Electrodes Other Electrodes
The electrode kit contains four voltage/current electrodes, plus four luer-tipped cartridges. Electrodes are threaded and screw securely into the end of each cartridge.
The luer tip then plugs securely into the luer openings of the chamber. The cable from each electrode terminates with a 2 mm pin which may be plugged into voltage/current clamps such as WPI’s DVC1000 or EVC-4000.
The miniature electrode-gel cartridge is a small plastic tube with a male luer tip identical to those at the tip of hypodermic syringes. The tube may be filled with different gel materials; agar is commonly used but other gel materials may also be satisfactory.