Rodent Guillotines
Supplier: WORLD PRECISION INSTRUMENTS LLC
DCAP-M DCAP DCAP-L
77664-196EA 1243 USD
77664-196 77664-192 77664-194
Rodent Guillotines
Knives
These guillotines are designed for small animal researchers, and they come in three sizes. The guillotine is considered one of the most humane methods of dispatching laboratory animals. With a single downward motion of the handle, you can euthanize a rodent quickly and without trauma.
  • Guillotine designed for small animal research
  • Three sizes for mice, rats and other small animals
  • Large, stable base
  • Hardened blades ensure you get many years of service and the blades remain sharp
  • Ambidextrous configuration allows comfortable use for right and left handed people
Benefits:
Magnetic force draws the blades together, ensuring a clean and precise cut through very strong bones and skin for a quick and humane animal termination
Spring action prevents blades from accidentally falling, safeguarding users
Fluoropolymer coated base makes cleanup easier
These guillotines are designed for small animal researchers, and they come in three sizes. The guillotine is considered one of the most humane methods of dispatching laboratory animals. With a single downward motion of the handle, you can euthanize a rodent quickly and without trauma.
The blades are comprised of hardened steel so they hold an edge and remain sharp, offering years of service. Sharp blades make the process swift for both the researcher and the subject. With proper care, the stainless steel blades resists corrosion, too.
A heavy duty spring at the top of each guillotine securely holds the blade in an open position. This prevents the blade from accidentally falling and injuring laboratory personnel. It also makes it quicker to dispatch the subject, requiring only a single downward action.
This guillotine was designed specifically for lab work. The large base affords greater stability. The long handle provides extra leverage. The spring action prevents the blades from falling unexpectedly. The hardened stainless blades ensures longevity of the unit. The simplified construction means easy maintenance. And the fluoropolymer-coated surface on the base makes cleaning easy. The DCAP guillotine is easy to use safely in your laboratory.
Benefits:
Blades are drawn together by magnetic force to ensure a clean and precise cut through very strong bones and skin
Spring action prevents blades from accidentally falling
Fluoropolymer coated base for easy cleanup
Applications:
Humane sacrifice of small animals