Quartz Cuvette F 100 mm, 28 ml
Supplier: WORLD PRECISION INSTRUMENTS LLC
CUV2071-1
77664-176EA 456 USD
77664-176
Quartz Cuvette F 100 mm, 28 ml
Cuvettes Spectrophotometer Cuvettes
Glass and synthetic quartz cuvettes are ideal for UV/VIS/NIR absorbance or fluorescence experiments. Synthetic quartz can be used in deep UV applications and is recommended for fluorescent applications, as it does not exhibit background fluorescence. Quartz cuvettes (absorbance, fluorescence and flow) are shipped individually packaged. These economic quartz are ideal for precision measurements because of their high quality materials used and their low manufacturing tolerances.
  • 102.5 L × 22 mm Ext.Ø
  • 28 ml V
  • Beam width 19 mm
Spectral Range (>80%): 200 to 2500 nm; Transmission Difference between different cuvettes: Less than 1% (Typical transmission curves of synthetic quartz cuvettes were measured empty, thickness 1.25 mm × 2, including surface reflections, measured with a TIDAS II against air as reference).