Calcium Buffers
Supplier: WORLD PRECISION INSTRUMENTS LLC
CALBUF-2
77664-082PK 275 USD
77664-082
Calcium Buffers
Buffers Assay Buffers
CALBUF-2 is especially suitable for calibrating fluorescent Ca⁺⁺ indicators. It provides eleven buffer standards in the 10⁻⁴ to 10⁻⁸ M Ca⁺⁺ range, whereas other commonly used fluorescent Ca⁺⁺ indicators have the apparent Kd in the range of 100 to 300 nM. As with any ionic sensitive indicator, the sensitivity range of these indicators is about 1.0 log unit above and below the Kd. CALBUF-2 provides seven calibration points in this sensitivity range. It has an osmolarity of 0.305, which is isotonic with most mammalian cells.
  • Use with calcium fluorescent indicators
  • 20 ml of each buffer
Concentrations: 1×10⁻⁸, 4×10⁻⁸, 2.1×10⁻⁷, 5×10⁻⁷, 7.5×10⁻⁷, 1×10⁻⁶, 4×10⁻⁶, 1×10⁻⁵, 4×10⁻⁵, 1×10⁻⁴ M at 20 °C. Ionic strength: 0.150 M. 11 bottles, 20 ml each.