Carbon Wire
Supplier: WORLD PRECISION INSTRUMENTS LLC
C3005
77664-078EA 82 USD
77664-078
Carbon Wire
Wire
Carbon wire is a single 30 µm fiber of electrochemically activated carbon. This fiber is especially useful in microelectrochemical experiments.
  • 49 AWG
  • 0.0012" (30 µm) Ø
  • 5' (1.5 m) long