AirTherm™ SMT Heat Controller/Humidity Monitors
Supplier: WORLD PRECISION INSTRUMENTS LLC
AIRTHERM-SMT-1W AIRTHERM-SMT-2W
77664-040EA 4995 USD
77664-040 77664-042
AirTherm™ SMT Heat Controller/Humidity Monitors
Controllers Temperature Controllers
The AirTherm™ SMT is a new generation of heat control system from WPI designed to be used in Live Cell Imaging applications with microscopes fitted with a full microscope environmental chamber enclosure. The standard AirTherm™ SMT controls temperature and, as an option, monitors humidity.
  • Precision heat controller for use in live cell imaging and custom incubators
  • Control heat and monitor humidity (optional) with a single controller
  • Electrically and acoustically quiet
  • Quick, precise response to thermal change
Benefits:
PID control algorithms allow for tight control of temperature in the environmental chamber
System includes hoses and probes so that it is ready to connect with your microscope chamber, incubator or controlled environment
Applications:
Live cell imaging systems fitted with full microscope environmental chamber enclosures
Control environment in custom incubators
Delivery information: P/N 5489; two connection hoses (4 foot) and four clamps are provided for a single inlet-outlet chamber (for dual inlet-outlet chambers refer to P/N 96824).