WPI’s filter holder for particulate absorption measurements is specially designed for field use. It is rugged and portable. It performs as well as a laboratory based spectrophotometer. It can be directly connected to WPI’s line of fiber optic spectrometers and light sources.
- Simple measurements for particulate absorption
- Rugged and portable
- Performs as well as a laboratory based spectrophotometer
Instead of collecting your samples, transporting them to a laboratory, and accepting the loss of spectral information associated with it (Sosik, 1999), particulate absorption can now be measured on site.
Application: Measurement of UV/VIS absorption of particulate matter in seawater
Benefits: QFT corrects for the effect of scattering; Averages out larger, non-organic particles