Metallurgy Objective
Supplier: WORLD PRECISION INSTRUMENTS LLC
800292
77663-976EA 661.27 USD
77663-976
Metallurgy Objective
Microscope Objectives
  • Long working distance objective for a microscope
  • 40× Magnification
  • 0.25 Numerical aperature