Mouse Rectal Probe Ball Tip
Supplier: WORLD PRECISION INSTRUMENTS LLC
61824
77663-918EA 394 USD
77663-918
Mouse Rectal Probe Ball Tip
Temperature Probes
The needle microprobes and animal rectal probes are all designed to withstand sterilization by ETO gas and gluteraldehyde solution. Our flexible implantable microprobes are both ETO gas sterilizable and autoclavable.
  • Rectal probe for use with the ATC2000, Animal temperature controller
  • The heating plate cable is 1.1 m long