Wire Eyelid Speculum
Supplier: WORLD PRECISION INSTRUMENTS LLC
555801L
77663-896EA 37 USD
77663-896
Wire Eyelid Speculum
Specula
A wire eyelid speculum is a surgical instrument used by ophthalmologists to keep eyelids open during an exam or surgery. It consists of two curved blades that are gently inserted between the upper and lower eyelids to hold them open, allowing the ophthalmologist a clear view of the eye.
  • 4.5 cm (1.75") long
  • 13.5 mm blades
  • Maximum spread: 20 mm
  • Non-magnetic and lightweight titanium
This wire speculum can be sterilized with a chemical sterilant or by autoclaving. Be sure to clean, disinfect and sterilize your surgical instruments promptly after each use to ensure they last for many years.