Silicone Handle and Knob Covers for PSMB5N/PSMT5N Surgical Microscope
Supplier: WORLD PRECISION INSTRUMENTS LLC
505126 505127 505129 505130
77663-836EA 24.66 USD
77663-836 77663-838 77663-840 77663-842
Silicone Handle and Knob Covers for PSMB5N/PSMT5N Surgical Microscope
Covers
Silicone handle covers for use with the SurgiScope Surgical Microscope. The silicone handle covers protect the handles and knobs of your microscope during surgery and can be easily removed for cleaning.
  • Knob covers and handle covers sold separately
  • Choose a set of 2 handle covers or a set of five knob covers