Luer to Tube Assortment Kit
Supplier: WORLD PRECISION INSTRUMENTS LLC
504954
77663-804EA 440.53 USD
77663-804
Luer to Tube Assortment Kit
Fittings and Connectors
This Luer coupler kit is useful for building your own liquid flow experiments. Luer coupler kit is handy for quick connects in polypropylene.
  • Over 250 assorted parts and luer couplers in each kit
  • Valves are polycarbonate, and the valve handles are polyethylene. Do not autoclave those parts
  • Polypropylene parts can be autoclaved repeatedly at 121 °C/15 psi., 15 min cycles
  • Polypropylene fittings are chemically inert and resistant to most organic and inorganic solvents
  • Nylon fittings are strong and can be bonded with adhesive
Assemble quick-disconnect luer fittings for use with flexible tubing with internal diameters of ¹/₁₆, ³/₃₂ and ¹/₈".
A variety of quick-disconnect connectors can be quickly made for connecting small diameter flexible tubing; 3-way connections can be made with the use of the 3-way luer tee; luer plugs, tees, connectors, bulk-head mounts, color coding rings, locking nuts, male and female luers - are all included to enhance the versatility of this kit.
The kit has over 250 assorted parts and is offered in two different types of materials. Nylon parts are not autoclavable. Polypropylene fittings are chemically inert and resistant to most organic and inorganic solvents.
Applications: Liquid flow experimental setups
Delivery information: The kit are over 250 assorted parts including barbed male Luers of various sizes with locking nuts or slip fits, female Luers for bulkhead mounting in several sizes, female Luer connectors with full Luer threads, threaded port male slip Luers, female Luer connector with full Luer threads, female Luer Tees with full Luer threads, threaded port male and female slip Luers of various sizes, barbed female Luers with full thread in various sizes, male Luer plugs with locking nuts, female Luer Plugs with full threads, locking male to female Luer check valves, male Luer plugs with locking nuts, female Luer plugs with full threads, locking male to female Luer check valves, locking male × female Luer 1-Way stopcock, locking male × locking male × female Luer 4-way stopcock, stamped hex nut for bulkhead Luers, locking male × female Luer 1-Way stopcock, locking male × locking male × female Luer 4-Way stopcock, stamped hex nut for bulkhead Luers, and coding rings with and without legs.