Swiss High Precision Tweezers, 12 cm
Supplier: WORLD PRECISION INSTRUMENTS LLC
504746 504747
77663-766EA 52.36 USD
77663-766 77663-768
Swiss High Precision Tweezers, 12 cm
Forceps
Swiss high precision tweezers are designed for tasks that require a high level of accuracy, delicacy, and control. These tweezers are commonly used in various industries and applications, including electronics, watchmaking, jewelry, microscopy, and medical procedures. The distinguishing features of high precision tweezers include their fine tips, and anti-magnetic or anti-static properties.
  • Anti-acid, anti-magnetic
  • Surgical grade stainless steel
  • Swiss made quality
Choose a thick pointed tip or a flat rounded tip. Fine pointed tips allow for precision handling of small or delicate objects. The flat rounded tips provide a broader and gentler surface for grasping delicate objects and give you an even pressure distribution.
For electronics work, the anti-magnetic steel prevents the tweezers from interfering with the magnetic components or causing damage to sensitive equipment. Both of these tweezers are quality Swiss-made instruments. These surgical instruments are autoclavable.