Stereotaxic Kit
Supplier: WORLD PRECISION INSTRUMENTS LLC
504743
77663-760EA 297.21 USD
77663-760
Stereotaxic Kit
Dissection Sets
Four piece sterotaxic surgical kit including popular instruments.
Delivery information: Kit includes: Scalpel handle #3, 13 cm, Metzenbaum scissors, 11.5 cm, SuperCut, straight, microdissecting forceps, 10.2 cm, angled, serrated, needle holder, 14 cm, round handles, straight.