Swiss High Temperature Coating (HTC) Thumb Tweezers
Supplier: WORLD PRECISION INSTRUMENTS LLC
504617 504619
77663-708EA 67.77 USD
77663-708 77663-710
Swiss High Temperature Coating (HTC) Thumb Tweezers
Forceps
These thumb tweezers have undergone a specialized coating process to withstand elevated temperatures with resistance up to 620 °C (1150 °F).
  • Curved blades with very fine, sharp tips or straight blades with flat tips
  • High corrosion and chemical resistance
  • Smooth, anti-glare surface
  • Swiss-made surgical grade stainless steel with HTC coating
The HTC coating can withstand exposure to elevated temperatures without compromising their structural integrity or performance. This is particularly useful in applications where the tweezers may be used in high-temperature environments, such as soldering or other industrial processes. The HTC coating provides enhanced corrosion resistance, protecting the tweezers from damage caused by exposure to heat and potentially corrosive substances. The coating also contributes to the overall durability of the tweezers, ensuring they maintain their functionality and appearance over time, even when subjected to high-temperature conditions. You get increased wear resistance, extending the lifespan of the tweezers and reducing the frequency of replacements.
These tweezers are 11 cm (4.75") long. Choose either straight tweezers with a blunt tip or a curved blade with fine, sharp tips. They have a smooth, non-glare surface. These thumb forceps are suitable for autoclaving or chemical sterilization.