Rapicide® OPA/28 Disinfectant
Supplier: WORLD PRECISION INSTRUMENTS LLC
504611
77663-698EA 84.7 USD
77663-698
Rapicide® OPA/28 Disinfectant
Cleaning Agents Disinfectants
Rapicide® OPA/28 high-level disinfectant: a superior way to reprocess devices that are unsuitable for sterilization. This allows the ultimate combination of safety, convenience and value.
  • For use as a high level disinfectant (HLD)
  • Suitable replacement for Cidex
  • Glutaraldehyde free
  • Low odor, effective and fast acting
  • Effective against glutaraldehyde resistant mycobacteria
  • Long lasting - up to 14 days resuse life
  • No activation or mixing required
  • Discard down healthcare drains according to local regulations
Protects workers by limiting exposure to disinfectants. Because of the low percentage of OPA and low vapor pressure, irritation to the eyes, nose and throat are significantly reduced, as compared to Glutaraldehyde.
With a 10-minute high-level manual disinfection time provides at least one extra disinfection cycle for every five instruments reprocessed. No mixing or activation required, comes ready to use. Excellent materials compatibility, non-corrosive to metal features.
Effectively inactivates TB, HIV2, MRSA, VRE and Hepatitis. Maximum reuse period of 28 days.
The test strips are intuitive and fast with a pass/fail reference color chart on the bottle and 3-second dip time and 90-second test strip read time.
2 year unopened shelf life and 75 days once opened.