Stage Micrometer, 3 Ranges
Supplier: WORLD PRECISION INSTRUMENTS LLC
504606
77663-696EA 243.31 USD
77663-696
Stage Micrometer, 3 Ranges
Micrometers
Stage micrometer, 1×3" slide with linear targets etched into the glass.
3 ranges: 10 mm / 100 to 0.1 mm (100 μm) scale, 10 mm / 200 to 0.05 mm (50 μm) scale and 1 mm / 100 to 0.01 mm (10 μm) small scale.