Small Syringe Multi-Rack for SPLG Syringe Pumps
Supplier: WORLD PRECISION INSTRUMENTS LLC
504576
77663-694EA 1997.64 USD
77663-694
Small Syringe Multi-Rack for SPLG Syringe Pumps
Racks Syringe Racks
The multi-rack system allows the syringe pump to hold and control multiple syringes simultaneously. This is particularly useful when there is a need for infusing multiple fluids or drugs into a system at different rates or volumes.
  • Ensure precise and synchronized delivery when doing continuous, simultaneous infusion of multiple solutions
  • The ability to use different syringe sizes and types in the same pump setup provides flexibility in experimental design
Each syringe in the rack can be independently controlled, enabling more complex experimental setups. This can save time and streamline experimental processes.
Researchers can choose syringes of varying capacities based on the specific requirements of each infusion. Save space by using one pump instead of multiple individual syringe pumps for different infusions. A multi-rack system consolidates these pumps into a single unit. While the initial cost of a multi-rack syringe pump may be higher than a single-channel pump, it can be more cost-effective when compared to purchasing multiple single-channel pumps for the same experimental setup.
Small syringe multi rack for six 30 - 60 ml syringes or 10 0.5 µl - 20 ml syringes.
Work with the SPLG200, SPLG210 or SPLG210P.