LiquiVet Rapid Tissue Adhesive
Supplier: WORLD PRECISION INSTRUMENTS LLC
504561
77663-686EA 40.05 USD
77663-686
LiquiVet Rapid Tissue Adhesive
Adhesives
LiquiVet monomeric formulation forms a thin, waterproof adhesion to bond tissue in a variety of vet procedures. Comes in 3 g dispensing bottles with five applicators.
  • Fast-acting cyanoacrylate-based tissue adhesive for wound care
  • Polymerizes in 2 seconds, ideal for skin closure
  • Superior performance in the time necessary to close a wound
Excellent for quick repair of minor lacerations, abrasions, and other minor wounds.
For external lacerations, not to be used internally.
Caution: Veterinary use only.