Westcott Spring Scissors with Angled Tips
Supplier: WORLD PRECISION INSTRUMENTS LLC
504492
77663-630EA 187.87 USD
77663-630
Westcott Spring Scissors with Angled Tips
Scissors
Westcott rounded-blade, side-angled spring scissors are popular spring handled scissors that automatically reopen after each cut. Westcott brand spring scissors give you an ergonomic and comfortable grip, reducing hand strain and fatigue during repetitive cutting tasks.
  • Made of surgical grade stainless steel and are suitable for autoclaving or chemical sterilization
  • Rounded blades with blunt tips
Westcott spring scissors are perfect for ambidextrous use, often preferred by left-handed individuals. The tips of these surgical scissors are 21 mm long and angled to the side, and they are available with blunt tips.