Student Anatomy Kit II
Supplier: WORLD PRECISION INSTRUMENTS LLC
504471
77663-590EA 83.16 USD
77663-590
Student Anatomy Kit II
Dissection Sets
Basic kit for anatomy studies.
Contains 18 pieces.
Delivery information: Kit contains: 1 Plastic ruler, 6"; 1 Teaser needle angled; 1 Teaser needle straight; 1 Dressing forceps, 16 cm; 1 Mosquito forceps, 12.5 cm; 1 Scalpel handle #3; 4 Blades for scalpel handle #3; 1 Scalpel handle #4; 4 Blades for Scalpel handle #4; 1 Dissecting scissors, 14 cm sharp/blunt; 1 Spencer scissors, 10.75 cm; 1 Leather pouch.