Mini Centrifuges 8 Place, 240 V
Supplier: WORLD PRECISION INSTRUMENTS LLC
504328
77663-584EA 754.33 USD
77663-584
Mini Centrifuges 8 Place, 240 V
Centrifuges Microcentrifuges
This personal centrifuge fits in the palm of your hand. It is deal for quick spin downs of either microtubes or PCR tubes. Close the lid and the rotor instantly ramps up to 6000 rpm. Open the lid and it decelerates to a stop.
  • Storace compartment for PCR rotor
  • Quiet operation
  • Starts and stops in seconds
  • One year warranty
The 8-position microtube rotor is incuded, along with a second rotor for 0.2 ml PCR tubes and strips. The second rotor is stored (with the removal key) in a compartment on the bottom of the centrifuge.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (77663-582) are also available, please see separate product page.