Fixing Screws
Supplier: WORLD PRECISION INSTRUMENTS LLC
504281
77663-572EA 21.58 USD
77663-572
Fixing Screws
Fasteners Screws
Fixing screw for securing the internal cannula to the guide cannula, to prevent movement during injection.
  • One size fits all