Quantrex® Ultrasonic Cleaner
Supplier: WORLD PRECISION INSTRUMENTS LLC
504216
77663-560EA 1408.98 USD
77663-560
Quantrex® Ultrasonic Cleaner
Ultrasonic Cleaners
Quantrex® bath with a timer.
  • Provides super-strength cleaning every time
  • When used with L&R’s specialty formulated solutions, the self-contained, compact unit offers efficient trouble-free cleaning
  • Each Quantrex machine comes standard with increased power—strength you can see as soon as you turn the unit on
  • Stainless steel drain with multi-positional outlet for easy removal of solution
  • 14 quality inspection steps for strength and durability
It is made of vinyl-stainless steel and stainless steel.
Application: Versatile enough for a variety of cleaning applications.
Delivery information: Includes timer, drain and cover and heater.