Barrier Milk Spray
Supplier: WORLD PRECISION INSTRUMENTS LLC
504215
77663-558EA 41.59 USD
77663-558
Barrier Milk Spray
Sprays
Barrier Milk Ready to Use Spray helps prevent corrosion of instruments when used prior to sterilization. Protects and extends the life of instruments during autoclaving.
  • Nitrate-free
  • Chromate-free
  • Phenolic-free
  • Non-irritating
  • Neutral pH
  • Mild, pleasant odor
  • Corrosion inhibitor
Simply spray the instrument directly before autoclaving with the unique water-based blend of lubricants and corrosion inhibitors. The fine mist works its way into hinges and box locks as well as covering all instruments with protection.
Caution: Use before autoclaving.