LED Ring Light
Supplier: WORLD PRECISION INSTRUMENTS LLC
504134
77663-500EA 426.55 USD
77663-500
LED Ring Light
Lights LED Lights, Lamps and Bulbs
'White' light illumination with 72 LED bulbs.
  • Maximum opening 61 mm
  • Brightness adjustable
  • ESD safe
  • Direct top illumination plus four zones for shadows
  • Low cost
  • Less bulky than halogen
  • Cold light with no heat
Ring light is divided into four areas and each area is turned on and off separately.
Application: Stereo microscope use