Rat Femoral Vein Catheter
Supplier: WORLD PRECISION INSTRUMENTS LLC
504119
77663-492EA 169.4 USD
77663-492
Rat Femoral Vein Catheter
Catheters
Catheters used for sampling or dosing can vary and depend on access or animal size. We carry catheters used for common applications.
  • Rat femoral vein catheter
  • 3F Lumen
  • Three bead
All catheters are ETO sterilized.