Magnetic Base
Supplier: WORLD PRECISION INSTRUMENTS LLC
503568
77663-332EA 341.86 USD
77663-332
Magnetic Base
Magnets
An ideal accessory for optical tables and vibration-free platform. Reduces experimental set-up time by allowing free positioning and instant clamp down of optical components. Switchable ON/OFF magnetic circuit permits fine adjustment and precise positioning.
  • Easy ON/OFF operation using lever
  • Thin and powerful magnetic force
  • Generous array of tap holes