Tweezer, Plastic
Supplier: WORLD PRECISION INSTRUMENTS LLC
503362
77663-286EA 9.25 USD
77663-286
Tweezer, Plastic
Forceps
Delrin plastic is an engineering thermoplastic used in precision parts requiring high stiffness, low friction and excellent dimensional stability.
  • High abrasion resistance
  • Low coefficient of friction
  • High heat resistance
  • Good electrical and dielectric properties
  • Low water absorption
  • Antistatic, antimagnetic, waterproof
  • Heat resistant to 240 °C
  • The FDA approved Delrin for use in the food industry
As with many other synthetic polymers, it is produced by different chemical firms with slightly different formulas and sold variously by such names as Delrin, Celcon, Duracon and Hostaform.