Mini Dissecting Scissors, Straight, 9.5 cm
Supplier: WORLD PRECISION INSTRUMENTS LLC
503241 503242 503243 503244 503245
77663-180EA 63.15 USD
77663-180 77663-182 77663-184 77663-186 77663-188
Mini Dissecting Scissors, Straight, 9.5 cm
Scissors
Our mini dissecting scissors with Large Rings are similar to regular dissecting scissors, but they are shorter and have more delicate tips, making them ideal for fine dissection work in tight spaces. They are commonly used in microsurgery, like ophthalmic surgery. These dissecting scissors have large rings for people with larger fingers.
  • These scissors are 9.5 cm (3.75") long
  • Straight blades with rounded tips and fine blades
These dissecting scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. (Standard and medium blades are also available on our mini dissecting scissors).
Dissecting scissors have thinner (and often longer) blades than operating scissors. They are designed for cutting and probing in tight spaces. Their tips are pointed, where operating scissors are generally blunt. These are available in surgical grade stainless steel.
These dissecting scissors are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved. To guarantee a long life for your surgical instruments, be sure to clean them thoroughly after each use.