Stainless Steel Clamps
Supplier: WORLD PRECISION INSTRUMENTS LLC
14073-4 503041 503078-4 503079-4 503080-4 503082-4 503086 503042
77661-698PK 15 USD
77661-698 77663-074 77663-096 77663-098 77663-100 77663-102 77663-110 77663-076
Stainless Steel Clamps
Clamps General Purpose Clamps
These high quality components are made of stainless steel and polymer that resists organic solvents and corrosion.
They can be easily assembled to make a stand-alone setup for student laboratory equipment or to make a complicated frame for research labs. Use these components to custom build your own stands for laboratory glassware, manipulator stands or any other laboratory application you can envision.