Standard Probe Holder
Supplier: WORLD PRECISION INSTRUMENTS LLC
502210
77663-068EA 611.34 USD
77663-068
Standard Probe Holder
Electrode Holders
Standard probe holder/corner mount.
  • For use with stereotaxic frames
  • Holds probes 0.3 to 1.5 mm O.D