Magnetize/Demagnetize Instrument
Supplier: WORLD PRECISION INSTRUMENTS LLC
501900
77662-944EA 73.94 USD
77662-944
Magnetize/Demagnetize Instrument
Magnets
Instantly magnetize or demagnetize small surgical instruments and small steel parts that are over 1.3" in length and 0.125" in diameter.
  • No batteries or electricity required
  • Box is high quality molded ABS plastic, impact resistant and cadmium free
  • The innovative step design lets you control the level of magnetism
To Magnetise a tool: Insert tool into the "+" opening and slide back and forth. Continue this procedure until the desired level of magnetism is reached.
To Demagnetise a tool: Insert tool at the lowest "−" opening. Slide back and forth.More Product Information