Surgical Table
Supplier: WORLD PRECISION INSTRUMENTS LLC
501892
77662-932EA 112.42 USD
77662-932
Surgical Table
Animal Supplies Animal Restraints
Constructed of clear acrylic, this economical surgery table features adjustable nylon cords for fast animal restraint and a slight incline for easy viewing.
  • 4 cm high in the rear
  • 2 cm high in the front
The acrylic construction is durable, yet easy to clean. The nylon cords may be adjusted at the point of contact with the animal and also under the table itself. The surgery table is designed for use in dissecting small animals such as frogs, birds, mice, rats, guinea pigs, etc.