Alm Self-Retaining Retractor
Supplier: WORLD PRECISION INSTRUMENTS LLC
501844
77662-880EA 100.11 USD
77662-880
Alm Self-Retaining Retractor
Retractors
Smaller, thinner prongs for use on mouse/rat eyelids.
  • 2×2 prongs
  • Maximum spread: 5.6 cm