Bone Wax
Supplier: WORLD PRECISION INSTRUMENTS LLC
501771
77662-834EA 53.9 USD
77662-834
Bone Wax
First Aid Supplies First Aid Kit Components
Achieve homeostasis of bone by applying a small amount of sterile bone wax. The wax is a combination of beeswax, isopropyl palmitate and mineral oil.
  • Sterile
  • Stops bleeding at the site