Crile Hemostatic Forceps
Supplier: WORLD PRECISION INSTRUMENTS LLC
501730 501730-G 501713-G 503227 503228 503230 501242 501712 501713
77662-742EA 58.52 USD
77662-742 77662-744 77662-696 77663-162 77663-164 77663-166 77662-476 77662-692 77662-694
Crile Hemostatic Forceps
Forceps
Crile hemostatic forceps are atraumatic and non-toothed clamps used to clamp blood vessels, grab tissue or vessels.
They are often known as a 'snap.'